Natrium-N-methyltaurinat 95%

Lieferant: Apollo Scientific
Achtung

OR943245-5G OR943245-25G OR943245-100G
APOSOR943245-5GEA 21.9 EUR
APOSOR943245-5G APOSOR943245-25G APOSOR943245-100G
Natrium-N-methyltaurinat 95%
Natrium-N-methyltaurinat

60% in water.

Formel: C₃H₈NNaO₃S
Dichte: 1,21
MDL: MFCD00066598
CAS-Nummer: 4316-74-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR