4-Bromo-2,3,6-trifluorophenol 95%

Lieferant: Apollo Scientific
Achtung

PC47872-250MG PC47872-1G PC47872-5G
APOSPC47872-250MGEA 116 EUR
APOSPC47872-250MG APOSPC47872-1G APOSPC47872-5G
4-Bromo-2,3,6-trifluorophenol 95%
4-Bromo-2,3,6-trifluorophenol
Formel: C₆H₂BrF₃O
Molecular Weight: 226.979 g/mol
MDL: MFCD22414509
CAS-Nummer: 192446-70-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR