8-Amino-1-naphthalinsulfonsäure 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
8-Aminonaphthalene-1-sulphonic acid
OR932808-5G
OR932808-25G
OR932808-100G
APOSOR932808-5GEA
21.9
EUR
APOSOR932808-5G
APOSOR932808-25G
APOSOR932808-100G
8-Amino-1-naphthalinsulfonsäure 95%
8-Amino-1-naphthalenesulphonic acid
Formel:
C₁₀H₉NO₃S Molecular Weight: 223.25 g/mol Schmelzpunkt: > 300 °C |
MDL:
MFCD00035730 CAS-Nummer: 82-75-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...