Ethyl-3-iodimidazo[1,2-a]pyridin-2-carboxylat 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Iodo-imidazo[1,2-a]pyridine-2-carboxylic acid ethyl ester, ethyl 3-iodoimidazo[1,2-a]pyridine-2-carboxylate

OR930103-250MG
APOSOR930103-250MGEA 78.3 EUR
APOSOR930103-250MG
Ethyl-3-iodimidazo[1,2-a]pyridin-2-carboxylat 95%
Ethyl-3-iodimidazo[1,2-a]pyridin-2-carboxylat
Formel: C₁₀H₉IN₂O₂
Siedepunkt: 144…146 °C
MDL: MFCD06739188
CAS-Nummer: 292858-07-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR