3-Bromo-2-chloro-5-hydrazinylpyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: (5-Bromo-6-chloropyridin-3-yl)hydrazine

OR100001-250MG OR100001-1G
APOSOR100001-250MGEA 174 EUR
APOSOR100001-250MG APOSOR100001-1G
3-Bromo-2-chloro-5-hydrazinylpyridine
3-Bromo-2-chloro-5-hydrazinylpyridine
Formel: C₅H₅BrClN₃
Molecular Weight: 222.47 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD33440211
CAS-Nummer: 2357973-88-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR