8-Bromoquinoline-4-carbonitrile 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
8-Bromo-4-cyanoquinoline, 8-Bromoquinoline-4-carbonitrile
OR70310-250MG
OR70310-1G
APOSOR70310-250MGEA
219
EUR
APOSOR70310-250MG
APOSOR70310-1G
8-Bromoquinoline-4-carbonitrile 95%
8-Bromoquinoline-4-carbonitrile
Formel:
C₁₀H₅BrN₂ Molecular Weight: 233,07 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD12024578 CAS-Nummer: 1190315-89-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...