4-Amino-2-(methylthio)pyrimidine-5-carbaldehyde 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Amino-2-methylsulphanylpyrimidine-5-carbaldehyde
OR918031-1G
OR918031-5G
OR918031-10G
APOSOR918031-1GEA
88.6
EUR
APOSOR918031-1G
APOSOR918031-5G
APOSOR918031-10G
4-Amino-2-(methylthio)pyrimidine-5-carbaldehyde 98%
4-Amino-2-(methylthio)pyrimidine-5-carbaldehyde
Formel:
C₆H₇N₃OS Molecular Weight: 169.2 g/mol Schmelzpunkt: 180…190 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08669438 CAS-Nummer: 770-31-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...