2-Phenylcyclobutan-1-one 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Phenylcyclobutanone

OR922765-250MG OR922765-500MG OR922765-1G
APOSOR922765-250MGEA 364 EUR
APOSOR922765-250MG APOSOR922765-500MG APOSOR922765-1G
2-Phenylcyclobutan-1-one 97%
2-Phenylcyclobutan-1-one
Formel: C₁₀H₁₀O
Molecular Weight: 146.19 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD19382274
CAS-Nummer: 42436-86-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR