Ethyl 4-aminothiazole-5-carboxylate 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
ethyl 4-amino-1,3-thiazole-5-carboxylate
OR942787-250MG
OR942787-1G
OR942787-5G
APOSOR942787-250MGEA
31.9
EUR
APOSOR942787-250MG
APOSOR942787-1G
APOSOR942787-5G
Ethyl 4-aminothiazole-5-carboxylate 97%
Ethyl 4-aminothiazole-5-carboxylate
Formel:
C₆H₈N₂O₂S Molecular Weight: 172.21 g/mol Siedepunkt: 300.7/760 mm °C Flammpunkt: 135.6 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD06798219 CAS-Nummer: 152300-59-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...