Indol-3-carbonitril

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Cyan-1H-indol, 3-Indolcarbonitril, 1H-Indol-3-carbonitril, 3-Cyanindol

OR10194-1G OR10194-5G
APOSOR10194-1GEA 21.9 EUR
APOSOR10194-1G APOSOR10194-5G
Indol-3-carbonitril
Indol-3-carbonitril
Formel: C₉H₆N₂
Schmelzpunkt: 179 - 182 °C
MDL: MFCD00022717
CAS-Nummer: 5457-28-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR