L-Histidine-β-naphthylamide

Lieferant: Apollo Scientific
Achtung

Synonyme: H-His-betaNA, H-His-ßNA

OR36362-1G OR36362-5G OR36362-250MG
APOSOR36362-1GEA 226 EUR
APOSOR36362-1G APOSOR36362-5G APOSOR36362-250MG
L-Histidine-β-naphthylamide
L-Histidine-beta-naphthylamide
Formel: C₁₆H₁₆N₄O
Molecular Weight: 280.331 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00051166
CAS-Nummer: 7424-15-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR