1-Bromo-4-(bromomethyl)-2-methoxybenzene 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-Bromo-3-methoxybenzyl bromide
OR400145-250MG
OR400145-1G
OR400145-5G
APOSOR400145-250MGEA
105
EUR
APOSOR400145-250MG
APOSOR400145-1G
APOSOR400145-5G
1-Bromo-4-(bromomethyl)-2-methoxybenzene 95%
1-Bromo-4-(bromomethyl)-2-methoxybenzene
Formel:
C₈H₈Br₂O Molecular Weight: 279.96 g/mol |
MDL:
MFCD17169084 CAS-Nummer: 1148110-16-0 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...