1,10-Diaminodecan
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Decamethylendiamin, 1,10-Decandiamin
OR51847-25G
OR51847-100G
OR51847-500G
APOSOR51847-25GEA
21.9
EUR
APOSOR51847-25G
APOSOR51847-100G
APOSOR51847-500G
1,10-Diaminodecan
1,10-Diaminodecan
Formel:
C₁₀H₂₄N₂ Molecular Weight: 172,31 g/mol Siedepunkt: 140/12 mm °C Schmelzpunkt: 59…61 °C |
MDL:
MFCD00008151 CAS-Nummer: 646-25-3 UN: 3259 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...