Ethyl 7-(trifluoromethoxy)-1-benzofuran-2-carboxylate

Lieferant: Apollo Scientific
Achtung

PC301061-1G PC301061-250MG
APOSPC301061-1GEA 451 EUR
APOSPC301061-1G APOSPC301061-250MG
Ethyl 7-(trifluoromethoxy)-1-benzofuran-2-carboxylate
Ethyl 7-(trifluoromethoxy)-1-benzofuran-2-carboxylate
Formel: C₁₂H₉F₃O₄
Molecular Weight: 274.193 g/mol
MDL: MFCD34596998
CAS-Nummer: 896732-44-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR