Methyl 5-bromo-2-(trifluoromethyl)isonicotinate 95%

Lieferant: Apollo Scientific
Achtung

PC421064-1G PC421064-5G PC421064-10G
APOSPC421064-1GEA 97.3 EUR
APOSPC421064-1G APOSPC421064-5G APOSPC421064-10G
Methyl 5-bromo-2-(trifluoromethyl)isonicotinate 95%
Methyl 5-bromo-2-(trifluoromethyl)isonicotinate
Formel: C₈H₅BrF₃NO₂
MDL: MFCD16657895
CAS-Nummer: 1211589-41-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR