Methyl 4-cyano-5-fluoro-2-methylbenzoate 95%

Lieferant: Apollo Scientific
Gefahr

PC57432-250MG PC57432-1G PC57432-5G
APOSPC57432-250MGEA 172 EUR
APOSPC57432-250MG APOSPC57432-1G APOSPC57432-5G
Methyl 4-cyano-5-fluoro-2-methylbenzoate 95%
Methyl 4-cyano-5-fluoro-2-methylbenzoate
Formel: C₁₀H₈FNO₂
Molecular Weight: 193.17 g/mol
MDL: MFCD23707556
CAS-Nummer: 1427377-85-2
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR