5-Amino-2,2-difluor-1,3-benzodioxol 94%
Lieferant: Apollo Scientific
|
Gefahr
|
PC99476-1G
PC99476-5G
PC99476-10G
APOSPC99476-1GEA
21.9
EUR
APOSPC99476-1G
APOSPC99476-5G
APOSPC99476-10G
5-Amino-2,2-difluor-1,3-benzodioxol 94%
5-Amino-2,2-difluor-1,3-benzodioxol
Formel:
C₇H₅F₂NO₂ Siedepunkt: 95…97 °C Dichte: 1,41 |
MDL:
MFCD00190144 CAS-Nummer: 1544-85-0 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...