3-Bromo-2,5,6-trifluorophenol 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC502417-250MG
PC502417-1G
PC502417-5G
APOSPC502417-250MGEA
208
EUR
APOSPC502417-250MG
APOSPC502417-1G
APOSPC502417-5G
3-Bromo-2,5,6-trifluorophenol 95%
3-Bromo-2,5,6-trifluorophenol
Formel:
C₆H₂BrF₃O Molecular Weight: 226,98 g/mol |
MDL:
MFCD30529443 CAS-Nummer: 1936039-79-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...