6-Bromo-4-fluorobenzo[b]thiophene-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

PC502533-1G PC502533-5G
APOSPC502533-1GEA 928 EUR
APOSPC502533-1G APOSPC502533-5G
6-Bromo-4-fluorobenzo[b]thiophene-2-carboxylic acid
6-Bromo-4-fluorobenzo[b]thiophene-2-carboxylic acid
Formel: C₉H₄BrFO₂S
MDL: MFCD07701147
CAS-Nummer: 826995-57-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR