Ethyl 2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
PC301029-250MG
PC301029-1G
APOSPC301029-250MGEA
139
EUR
APOSPC301029-250MG
APOSPC301029-1G
Ethyl 2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Ethyl 2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Formel:
C₁₁H₁₀F₂N₂O₂ |
MDL:
MFCD32689358 CAS-Nummer: 2451256-48-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...