S-(Difluoromethyl) benzenesulphonothioate 95%

Lieferant: Apollo Scientific
Gefahr

PC502992-250MG PC502992-1G
APOSPC502992-250MGEA 219 EUR
APOSPC502992-250MG APOSPC502992-1G
S-(Difluoromethyl) benzenesulphonothioate 95%
S-(Difluoromethyl) benzenesulphonothioate
Formel: C₇H₆F₂O₂S₂
MDL: MFCD32070719
CAS-Nummer: 2022186-75-8
UN: 3265

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR