2-Brom-4-(trifluormethoxy)benzonitril 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC57115-1G
PC57115-5G
PC57115-10G
APOSPC57115-1GEA
21.9
EUR
APOSPC57115-1G
APOSPC57115-5G
APOSPC57115-10G
2-Brom-4-(trifluormethoxy)benzonitril 97%
2-Brom-4-(trifluormethoxy)benzonitril
Formel:
C₈H₃BrF₃NO Molecular Weight: 266,0147 g/mol |
MDL:
MFCD13185874 CAS-Nummer: 1214334-83-4 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...