3’-Chloro-5’-iodo-2,2,2-trifluoroacetophenone 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(3-Chloro-5-iodophenyl)-2,2,2-trifluoroethanone

PC57416-250MG PC57416-1G PC57416-5G
APOSPC57416-250MGEA 171 EUR
APOSPC57416-250MG APOSPC57416-1G APOSPC57416-5G
3’-Chloro-5’-iodo-2,2,2-trifluoroacetophenone 95%
3’-Chloro-5’-iodo-2,2,2-trifluoroacetophenone
Formel: C₈H₃ClF₃IO
Molecular Weight: 334.46 g/mol
MDL: MFCD33566604
CAS-Nummer: 2570109-75-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR