4-Bromo-2-(difluoromethyl)benzaldehyde 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC57736-250MG
PC57736-1G
PC57736-5G
APOSPC57736-250MGEA
190
EUR
APOSPC57736-250MG
APOSPC57736-1G
APOSPC57736-5G
4-Bromo-2-(difluoromethyl)benzaldehyde 95%
4-Bromo-2-(difluoromethyl)benzaldehyde
Formel:
C₈H₅BrF₂O Molecular Weight: 235.03 g/mol |
MDL:
MFCD32070692 CAS-Nummer: 2676196-10-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...