7-Bromo-5-(trifluoromethoxy)-1H-indazole
Lieferant: Apollo Scientific
|
Achtung
|
PC57417-250MG
PC57417-1G
APOSPC57417-250MGEA
219
EUR
APOSPC57417-250MG
APOSPC57417-1G
7-Bromo-5-(trifluoromethoxy)-1H-indazole
7-Bromo-5-(trifluoromethoxy)-1H-indazole
Formel:
C₈H₄BrF₃N₂O |
MDL:
MFCD34169326 CAS-Nummer: 2676196-05-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...