Sodium 2,5-difluorobenzoate
Lieferant: Apollo Scientific
Achtung
Synonyme: 2,5-Difluorobenzoic acid sodium salt
PC6585-50G PC6585-250G PC6585-1KG
APOSPC6585-50GEA 79.9 EUR
APOSPC6585-50G APOSPC6585-250G APOSPC6585-1KG
Sodium 2,5-difluorobenzoate
Sodium 2,5-difluorobenzoate
Formel: C₇H₃F₂NaO₂
Molecular Weight: 180,08 g/mol
MDL: MFCD03094396
CAS-Nummer: 522651-42-9