2-Dicyclohexylphosphino-2',6'-diisopropoxybiphenyl

Lieferant: Apollo Scientific
Achtung

OR922023-5G OR922023-25G OR922023-100G
APOSOR922023-5GEA 24.4 EUR
APOSOR922023-5G APOSOR922023-25G APOSOR922023-100G
2-Dicyclohexylphosphino-2',6'-diisopropoxybiphenyl
2-Dicyclohexylphosphino-2',6'-diisopropoxybiphenyl
Formel: C₃₀H₄₃O₂P
Schmelzpunkt: 123…126 °C
MDL: MFCD06798294
CAS-Nummer: 787618-22-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR