4-Acetyl-1-naphthoic acid 97%
Lieferant: Apollo Scientific
Achtung
OR31056-1G OR31056-5G OR31056-10G
APOSOR31056-1GEA 59.6 EUR
APOSOR31056-1G APOSOR31056-5G APOSOR31056-10G
4-Acetyl-1-naphthoic acid 97%
4-Acetyl-1-naphthoic acid
Formel: C₁₃H₁₀O₃
Molecular Weight: 214,22 g/mol
MDL: MFCD24480081
CAS-Nummer: 131986-05-5