4-Amino-2,5-difluorobenzoic acid 95%

Lieferant: Apollo Scientific
Achtung

PC902764-1G PC902764-5G PC902764-25G
APOSPC902764-1GEA 52.2 EUR
APOSPC902764-1G APOSPC902764-5G APOSPC902764-25G
4-Amino-2,5-difluorobenzoic acid 95%
4-Amino-2,5-difluorobenzoic acid
Formel: C₇H₅F₂NO₂
Molecular Weight: 173.12 g/mol
MDL: MFCD06208208
CAS-Nummer: 773108-64-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR