5-Bromo-7-chlorobenzoxazole 95%

Lieferant: Apollo Scientific
Achtung

OR400936-250MG OR400936-1G
APOSOR400936-250MGEA 273 EUR
APOSOR400936-250MG APOSOR400936-1G
5-Bromo-7-chlorobenzoxazole 95%
5-Bromo-7-chlorobenzoxazole
Formel: C₇H₃BrClNO
Molecular Weight: 232.46 g/mol
MDL: MFCD16468585
CAS-Nummer: 1226072-26-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR