Hämatoxylin Hydrat, (max. 6% H₂O)

Lieferant: Thermo Scientific

Synonyme: Naturschwarz 1Hydroxybrasilin Hydrat(+)-Hämatoxylin Hydrat(6aS,11bR)-7,11b-Dihydro-6H-indeno[2,1-c]chromen-3,4,6a,9,10-pentol Hydrat

A12431.14EA 96.5 EUR
A12431.14 A12431.06 A12431.22
Hämatoxylin Hydrat, (max. 6% H₂O)
Hämatoxylin Hydrat
Formel: C₁₆H₁₄O₆·xH₂O
Molecular Weight: 302,29 g/mol (anhy)
Schmelzpunkt: 100…120 °C
MDL: MFCD00078111
CAS-Nummer: 517-28-2
EINECS: 208-237-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR