D(+)-Prolin

Lieferant: BACHEM BIOCHEMICA

4031089.0005 4031089.0100
BACE4031089.0005EA 0 EUR
BACE4031089.0005 BACE4031089.0100
D(+)-Prolin
D(+)-Prolin
Formel: C₅H₉NO₂
Schmelzpunkt: 223 °C
MDL: MFCD00064317
CAS-Nummer: 344-25-2
EINECS: 206-452-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR