D(+)-Prolin

Lieferant: FLUOROCHEM

011849-50G 011849-100G 011849-5G
FLUO011849-50GEA 0 EUR
FLUO011849-50G FLUO011849-100G FLUO011849-5G
D(+)-Prolin
D(+)-Prolin
Formel: C₅H₉NO₂
Schmelzpunkt: 223 °C
MDL: MFCD00064317
CAS-Nummer: 344-25-2
EINECS: 206-452-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR