(S)-(+)-1-Phenyl-1,2-ethandiol ≥97%
Lieferant: Thermo Scientific
Synonyme:
(S)-(+)-α,β-Dihydroxyethylbenzol, (S)-(+)-(1,2-Dihydroxyethyl)benzol, (S)-(+)-Phenylethylenglycol, (S)-(+)-Phenylethan-1,2-diol
L09836.03
L09836.MD
L09836.03EA
55.5
EUR
L09836.03
L09836.MD
(S)-(+)-1-Phenyl-1,2-ethandiol ≥97%
(S)-(+)-1-Phenyl-1,2-ethandiol
Purity: 97%
CAS No.: 25779-13-9
Molecular Formula: C8H10O2
Formula Weight: 138.17
Melting Point: 64-66°
Boiling Point: 272-274°
MDL No.: MFCD00066256
Formel:
C₈H₁₀O₂ Molecular Weight: 138,17 g/mol Siedepunkt: 272…274 °C Schmelzpunkt: 64…66 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066256 CAS-Nummer: 25779-13-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...