Hämatoxylin, Supelco®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
Synonyme:
Hydroxybrasilin, Naturschwarz 1
H9627-100G
H9627-25G
H9627-100GEA
570
EUR
H9627-100G
H9627-25G
Hämatoxylin, Supelco®
Hämatoxylin
Hematoxylin has been used for staining gastric cancer tissues, myocardial tissue, heart sections and ovarian nuclei.
- Hematoxylin is an organic stain which chelates with metal ions and binds to DNA and RNA
- It is used for staining nucleus of the cells
- Hematoxylin along with eosin stain is used in the diagnosis of invasion stage in tumors
Formel:
C₁₆H₁₄O₆ Molecular Weight: 302,28 g/mol Schmelzpunkt: 200 °C (Zers., Lit.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00078111 CAS-Nummer: 517-28-2 EINECS: 208-237-3 Merck Index: 13,04654 |
Spezifikation Testergebnisse
Appearance (Colour) | Faint Yellow to Yellow and Faint Orange to Orange and Faint Beige to Beige and Faint Brown to Brown |
Appearance (Form) | Powder |
Solubility (Colour) | Yellow to Orange to Brown |
Solubility (Turbidity) 1 mg/ml, EtOH |
Clear to Very Slightly Hazy |
UV Absorbance Lambda Max Approx. 292 NM, MeOH |
Pass |
Recommended Retest Period | 2 years |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...