Tartrazine ≥85% (Farbstoffgehalt), orangenes Pulver
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
FD and C Yellow No. 5, Filter Yellow, Acid Yellow 23, Sodium 5-oxo-1-(4-sulphonatophenyl)-4-((4-sulphonatophenyl)diazenyl)-4,5-dihydro-1H-pyrazole-3-carboxylate
A17682.14
A17682.22
A17682.36
A17682.14EA
20.1
EUR
A17682.14
A17682.22
A17682.36
Tartrazine ≥85% (Farbstoffgehalt), orangenes Pulver
Tartrazine
CAS No.: 1934-21-0
Molecular Formula: C16H9N4Na3O9S2
Formula Weight: 534.37
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: Orange
MDL No.: MFCD00148908
Formel:
C₁₆H₁₂N₄O₉S₂ Molecular Weight: 534,36 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00148908 CAS-Nummer: 1934-21-0 EINECS: 217-699-5 Merck Index: 14,9072 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...