Allyltrimethoxysilan

Lieferant: abcr

Synonyme: Trimethoxyallylsilane

AB108967.0050 AB108967.0500
ABCRAB108967.0050EA 0 EUR
ABCRAB108967.0050 ABCRAB108967.0500
Allyltrimethoxysilan
Allyltrimethoxysilan
Formel: H₂CCHCH₂Si(OCH₃)₃
Molecular Weight: 162.26 g/mol
MDL: MFCD00053865
CAS-Nummer: 2551-83-9
EINECS: 219-855-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR