3-Acetylindol 97%

Lieferant: Thermo Fisher Scientific

Synonyme: Indol-3-ylmethylketon, 3-Indolylmethylketon, 3-Indolyl methyl ketone, 1-(3-Indolyl)-1-ethanon

102590050 102590250
ACRO102590050EA 36 EUR
ACRO102590050 ACRO102590250
3-Acetylindol 97%
3-Acetylindol
Formel: C₁₀H₉NO
Molecular Weight: 159.19 g/mol
Siedepunkt: 144…145 °C (10 mmHg)
Schmelzpunkt: 185…191 °C
MDL: MFCD00005626
CAS-Nummer: 703-80-0
EINECS: 211-875-5

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to light brown Powder
Infrared spectrum Conforms
Melting point 185 to 191 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR