2-Amino-4,6-dihydroxypyrimidin 98%

Lieferant: Thermo Fisher Scientific

Synonyme: 2-Amino-6-hydroxy-1H-pyrimidin-4-on

103510250 103511000
ACRO103510250EA 45.8 EUR
ACRO103510250 ACRO103511000
2-Amino-4,6-dihydroxypyrimidin 98%
2-Amino-4,6-dihydroxypyrimidine
Formel: C₄H₅N₃O₂
Siedepunkt: 394 °C (1013 hPa)
Schmelzpunkt: >300 °C
MDL: MFCD00006094
CAS-Nummer: 56-09-7
EINECS: 200-256-5

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to light yellow to light pink Powder
Infrared spectrum Conforms
Titration excess NaOH ≥97.5 %
UV Lambda max 628 to 632 nm
Absorption Ratio P-15/P+15 0.94 to 1.14
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR