6-Aminouracil 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Amino-2,6-dihydroxypyrimidin, 4-Aminouracil, 6-Amino-2,4-dihydroxypyrimidin

103521000 103525000 103520250
ACRO103521000EA 44.6 EUR
ACRO103521000 ACRO103525000 ACRO103520250
6-Aminouracil 98%
6-Aminouracil
Formel: C₄H₅N₃O₂
Molecular Weight: 127.10 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006071
CAS-Nummer: 873-83-6
EINECS: 212-854-3

Bestellen

Spezifikation Testergebnisse

Appearance Cream to light brown Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific activity ≥1200 GDU/g
Residue after ignition ≤5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR