5-Amino-2-methoxypyridin 90% tech.
Lieferant: Thermo Scientific
|
Achtung
|
103950050
103950250
103951000
ACRO103950050EA
35.9
EUR
ACRO103950050
ACRO103950250
ACRO103951000
5-Amino-2-methoxypyridin 90% tech.
5-Amino-2-methoxypyridin
Formel:
C₆H₈N₂O Molecular Weight: 124,14 g/mol Siedepunkt: 85…90 °C (1 mmHg) Schmelzpunkt: 29…31 °C Dichte: 1,575 g/cm³ Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006264 CAS-Nummer: 6628-77-9 EINECS: 229-612-8 |
Spezifikation Testergebnisse
Appearance | Red to yellow-brown Liquid |
Infrared spectrum | Conforms |
GC | ≥88 % |
Clarity of solution | Passes test |
Specific optical rotation | +103° to +108° (20°C, 589 nm) (c=5, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...