4,4'-Diaminodiphenylether 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: p-Aminophenylether, 4,4'-Diaminodiphenylether, Bis(4-aminophenyl)ether

104330250 104331000 104335000
ACRO104330250EA 31.8 EUR
ACRO104330250 ACRO104331000 ACRO104335000
4,4'-Diaminodiphenylether 98%
4,4'-Diaminodiphenylether
Formel: C₁₂H₁₂N₂O
Molecular Weight: 200.24 g/mol
Siedepunkt: 190 °C (.1 mmHg)
Schmelzpunkt: 189…193 °C
Flammpunkt: 218 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007863
CAS-Nummer: 101-80-4
EINECS: 202-977-0
UN: 2811
ADR: 6.1 II

Bestellen

Spezifikation Testergebnisse

Appearance Beige Crystalline powder
Infrared spectrum Conforms
Melting point 189 to 193 °C
Titration with HClO4 ≥97.5 %
Iron (Fe) ≤500 ppm
Magnesium (Mg) ≤5000 ppm
Potassium (K) ≤500 ppm
Sodium (Na) ≤500 ppm
Strontium (Sr) ≤500 ppm
Heavy metals (ICP-OES) ≤30 ppm
Sulfur compounds (as SO4) ≤0.1 %
Insoluble matter ≤0.03 % (in HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR