Benzopinakol 98%
Lieferant: Thermo Scientific
Synonyme:
Tetraphenylethylenglycol, 1,1,2,2-Tetraphenyl-1,2-ethandiol
105590100
105591000
ACRO105590100EA
27.2
EUR
ACRO105590100
ACRO105591000
Benzopinakol 98%
Benzopinakol
Formel:
(C₆H₅)₂C(OH)C(OH)(C₆H₅)₂ Molecular Weight: 366,46 g/mol Siedepunkt: 508 °C (1013 hPa) Schmelzpunkt: 184…186 °C Dichte: 1,24 g/cm³ (20 °C) |
MDL:
MFCD00004448 CAS-Nummer: 464-72-2 EINECS: 207-356-8 Merck Index: 13,01101 |
Spezifikation Testergebnisse
Appearance | White to light yellow Powder or crystals or granules |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...