Benzoxazol 97+%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1,3-Benzoxazole

105680250 105681000 105680050
ACRO105680250EA 53.9 EUR
ACRO105680250 ACRO105681000 ACRO105680050
Benzoxazol 97+%
Benzoxazole
Formel: C₇H₅NO
Molecular Weight: 119.12 g/mol
Siedepunkt: 182…185 °C (1013 hPa)
Schmelzpunkt: 27…30 °C
Flammpunkt: 58 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00005765
CAS-Nummer: 273-53-0
EINECS: 205-988-9
UN: 1325
ADR: 4.1 III

Bestellen

Spezifikation Testergebnisse

Appearance Colorless to white to yellow Low melting solid or liquid
Infrared spectrum Conforms
Melting point 27 to 30 °C
GC ≥97.0 %
UV at 280 nm A: ≤0.01
Titration after Ion exchange ≥98.5 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR