5-Benzyloxyindol 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Benzyloxy-1H-indol

106090050 106090250
ACRO106090050EA 41.1 EUR
ACRO106090050 ACRO106090250
5-Benzyloxyindol 95%
5-Benzyloxyindol
Formel: C₁₅H₁₃NO
Molecular Weight: 223,27 g/mol
Siedepunkt: 165 °C (0,1 mmHg)
Schmelzpunkt: 96…102 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005676
CAS-Nummer: 1215-59-4
EINECS: 214-930-1
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to beige Powder
Infrared spectrum Conforms
GC ≤6 % Toluene
UV at 235 nm A: ≤1.0
Color scale ≤10 APHA
Stabilizer Approx. 50 ppm Amylene
Residue after evaporation ≤0.001 %
Acidity ≤0.0002 meq/g
Alkalinity ≤0.0002 meq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR