N-(2-Bromethyl)phthalimid 97%
Lieferant: Thermo Scientific
Synonyme:
N-(2-Bromethyl)phthalsäureimid, N-(2-Bromethyl)-1,2-benzoldicarbonsäureimid
106970250
106971000
ACRO106970250EA
33.5
EUR
ACRO106970250
ACRO106971000
N-(2-Bromethyl)phthalimid 97%
N-(2-Bromethyl)phthalimid
Formel:
C₁₀H₈BrNO₂ Molecular Weight: 254,08 g/mol Siedepunkt: 318 °C (1013 hPa) Schmelzpunkt: 81…84 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005902 CAS-Nummer: 574-98-1 EINECS: 209-379-9 |
Spezifikation Testergebnisse
Appearance | White to beige to light pink Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 81 to 84 °C |
Water | ≤1 % (K.F.) |
HPLC | ≥96.0 % (on dry substance) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...