5-Hydroxy-3-indolylessigsäure 99%

Lieferant: Thermo Scientific

Synonyme: 5-Hydroxyindol-3-essigsäure, 5-Hydroxyindol-3-ylessigsäure

121280010 121282500 121280050
ACRO121280010EA 202 EUR
ACRO121280010 ACRO121282500 ACRO121280050
5-Hydroxy-3-indolylessigsäure 99%
5-Hydroxy-3-indolylessigsäure
Formel: C₁₀H₉NO₃
Molecular Weight: 191,19 g/mol
Schmelzpunkt: 162…165 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00005639
CAS-Nummer: 54-16-0
EINECS: 200-195-4

Bestellen

Spezifikation Testergebnisse

Appearance Beige Crystalline powder
Infrared spectrum Conforms
Melting point 162 to 165 °C
Water ≤0.5 % (K.F.)
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR