2-Aminobenzothiazol 97%
Lieferant: Thermo Fisher Scientific
148810050
148811000
148815000
ACRO148810050EA
24.4
EUR
ACRO148810050
ACRO148811000
ACRO148815000
2-Aminobenzothiazol 97%
2-Aminobenzothiazole
Formel:
C₇H₆N₂S Siedepunkt: 190…195 °C (0,05 mmHg) Schmelzpunkt: 129…134 °C |
MDL:
MFCD00005785 CAS-Nummer: 136-95-8 EINECS: 205-268-4 Merck Index: 13,00423 |
Spezifikation Testergebnisse
Appearance | Beige to grey Powder or flakes |
Infrared spectrum | Conforms |
Melting point | 129 to 134 °C |
Water | ≤1 % |
HPLC | ≥97.0 % |
Titration with HClO4 | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...