2-Thioxo-2,3-dihydro-4(1H)-pyrimidinon 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Hydroxy-2-mercaptopyrimidin

151840050 151841000 151845000
ACRO151840050EA 21.3 EUR
ACRO151840050 ACRO151841000 ACRO151845000
2-Thioxo-2,3-dihydro-4(1H)-pyrimidinon 99%
2-Thiouracil
Formel: C₄H₄N₂OS
Molecular Weight: 128.15 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006039
CAS-Nummer: 141-90-2
EINECS: 205-508-8
UN: 3077
Merck Index: 12,09504

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C, 3 h)
HPLC ≥98.0 %
Appearance of solution (0.5% in 1M NaOH) Clear to light hazy
Titration with NaOH ≥98.5 %
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR