Uracil 99+%

Lieferant: Thermo Fisher Scientific

Synonyme: 2,4-Pyrimidindion, 2,4-Dihydroxypyrimidin, Pyrimidin-2,4(1H,3H)-dion

157300250 157301000 157300025 157305000
ACRO157300250EA 78.3 EUR
ACRO157300250 ACRO157301000 ACRO157300025 ACRO157305000
Uracil 99+%
Uracil
Formel: C₄H₄N₂O₂
Molecular Weight: 112.09 g/mol
Siedepunkt: 379
Schmelzpunkt: 335 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006016
CAS-Nummer: 66-22-8
EINECS: 200-621-9
Merck Index: 12,09985

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (115°C, 4 h)
Sulfated ash ≤0.3 %
HPLC ≥99.0 %
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR